Thermo Scientific Chemicals

2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals

Catalog number: 159491000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals

Catalog number: 159491000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 159491000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2295-31-0
IUPAC Name
1,3-thiazolidine-2,4-dione
Molecular Formula
C3H3NO2S
InChI Key
ZOBPZXTWZATXDG-UHFFFAOYSA-N
SMILES
O=C1CSC(=O)N1
Appearance (Color)
Light yellow
Appearance (Form)
Crystalline powder
Melting point
123°C to 126°C
HPLC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text