Informational:This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

KingFisher™ mL Food Protection Purification System Tube Rack

Catalog number: 15952
Have Questions?
Applied Biosystems™

KingFisher™ mL Food Protection Purification System Tube Rack

Catalog number: 15952
Have Questions?
Catalog Number
15952
also known as 159-52
Unit Size
1 unit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
15952
also known as 159-52
1 unit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Tube-sample rack, designed of use with the KingFisher mL Food Protection Purification System. The Tube Rack contains fifteen separate sample tube slots. Tubes for sample processing on the KingFisher mL Food Protection Purification System available separately (Cat. No. 159-51).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
KingFisher mL Food Protection Purification System
Product Line
KingFisher mL
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text