Dimethyl malonate, 97%, Thermo Scientific Chemicals
Dimethyl malonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl malonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 159562500
Price (USD)/ Each
30.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.60 
Save 4.95 (14%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
30.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.60 
Save 4.95 (14%)
Add to cart
Dimethyl malonate, 97%, Thermo Scientific Chemicals
Catalog number159562500
Price (USD)/ Each
30.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
35.60 
Save 4.95 (14%)
-
Add to cart
Chemical Identifiers
CAS108-59-8
IUPAC Name1,3-dimethyl propanedioate
Molecular FormulaC5H8O4
InChI KeyBEPAFCGSDWSTEL-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only