2,3-Diaminonaphthalene, 97%, Thermo Scientific Chemicals
2,3-Diaminonaphthalene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 159590250
Price (USD)/ Each
546.65
Online exclusive
643.00 
Save 96.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
546.65
Online exclusive
643.00 
Save 96.35 (15%)
Add to cart
2,3-Diaminonaphthalene, 97%, Thermo Scientific Chemicals
Catalog number159590250
Price (USD)/ Each
546.65
Online exclusive
643.00 
Save 96.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namenaphthalene-2,3-diamine
Molecular FormulaC10H10N2
InChI KeyXTBLDMQMUSHDEN-UHFFFAOYSA-N
SMILESNC1=C(N)C=C2C=CC=CC2=C1
Molecular Weight (g/mol)158.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Green-brown to brown
Melting point193°C to 199°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only