Thermo Scientific Chemicals

N-(3'-Aminopropyl)-2-pyrrolidinone, 95%, Thermo Scientific Chemicals

Catalog number: 159600050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(3'-Aminopropyl)-2-pyrrolidinone, 95%, Thermo Scientific Chemicals

Catalog number: 159600050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 159600050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7663-77-6
IUPAC Name
1-(3-aminopropyl)pyrrolidin-2-one
Molecular Formula
C7H14N2O
InChI Key
HJORCZCMNWLHMB-UHFFFAOYSA-N
SMILES
NCCCN1CCCC1=O
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms
GC
=<3 % 2-Pyrrolidinone
Appearance (Form)
Liquid
Refractive index
1.4970 to 1.4990 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text