Thermo Scientific Chemicals

1,5-Diazabicyclo[4.3.0]non-5-ene, 98%, Thermo Scientific Chemicals

Catalog number: 159611000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,5-Diazabicyclo[4.3.0]non-5-ene, 98%, Thermo Scientific Chemicals

Catalog number: 159611000
100 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 159611000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3001-72-7
IUPAC Name
2H,3H,4H,6H,7H,8H-pyrrolo[1,2-a]pyrimidine
Molecular Formula
C7H12N2
InChI Key
SGUVLZREKBPKCE-UHFFFAOYSA-N
SMILES
C1CN2CCCN=C2C1
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
GC
>=97.5 %
Refractive index
1.5190 to 1.5210 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text