2-Methyl-3-buten-2-ol, 97%
2-Methyl-3-buten-2-ol, 97%
2-Methyl-3-buten-2-ol, 97%
2-Methyl-3-buten-2-ol, 97%
Thermo Scientific Chemicals

2-Methyl-3-buten-2-ol, 97%

CAS: 115-18-4 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 159670010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS115-18-4
IUPAC Name2-methylbut-3-en-2-ol
Molecular FormulaC5H10O
InChI KeyHNVRRHSXBLFLIG-UHFFFAOYSA-N
SMILESCC(C)(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link