4-Methoxybenzyl alcohol, 98%
4-Methoxybenzyl alcohol, 98%
4-Methoxybenzyl alcohol, 98%
4-Methoxybenzyl alcohol, 98%
Thermo Scientific Chemicals

4-Methoxybenzyl alcohol, 98%

CAS: 105-13-5 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 159731000
Price (USD)
42.40
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.40
Each
Add to cart
Chemical Identifiers
CAS105-13-5
IUPAC Name(4-methoxyphenyl)methanol
Molecular FormulaC8H10O2
InChI KeyMSHFRERJPWKJFX-UHFFFAOYSA-N
SMILESCOC1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(25°C) 1.111 to 1.115
Appearance (Form)Liquid after melting
GC>=97.5 %
Refractive index1.5440 to 1.5470 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2538410Certificate of AnalysisMay 24, 2025159735000
2531446Certificate of AnalysisMay 24, 2025159735000
A0441620Certificate of AnalysisMay 17, 2025159735000, 159731000
2352478Certificate of AnalysisJan 29, 2025159735000
2350589Certificate of AnalysisJan 29, 2025159735000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection