Thermo Scientific Chemicals

5-Nonanone, 98%, Thermo Scientific Chemicals

Catalog number: 159761000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

5-Nonanone, 98%, Thermo Scientific Chemicals

Catalog number: 159761000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 159761000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
502-56-7
IUPAC Name
nonan-5-one
Molecular Formula
C9H18O
InChI Key
WSGCRAOTEDLMFQ-UHFFFAOYSA-N
SMILES
CCCCC(=O)CCCC
GC
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.4180 to 1.4200 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text