5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromosalicylaldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 159911000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1761-61-1
IUPAC Name5-bromo-2-hydroxybenzaldehyde
Molecular FormulaC7H5BrO2
InChI KeyMKKSTJKBKNCMRV-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to yellow to light beige
Infrared spectrumConforms
Melting point103°C to 107°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only