Thermo Scientific Chemicals

5-Methylfurfural, 98+%, Thermo Scientific Chemicals

Catalog number: 159921000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Methylfurfural, 98+%, Thermo Scientific Chemicals

Catalog number: 159921000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 159921000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
620-02-0
IUPAC Name
5-methylfuran-2-carbaldehyde
Molecular Formula
C6H6O2
InChI Key
OUDFNZMQXZILJD-UHFFFAOYSA-N
SMILES
CC1=CC=C(O1)C=O
GC
>=98.0 %
Refractive index
1.5290 to 1.5320 (20°C, 589 nm)
Appearance (Color)
Clear yellow to brown
Appearance (Form)
Liquid
Acid value
=<10 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text