Thermo Scientific Chemicals

2,4,6-Tribromophenol, 98%, Thermo Scientific Chemicals

Catalog number: 160105000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

2,4,6-Tribromophenol, 98%, Thermo Scientific Chemicals

Catalog number: 160105000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 160105000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
118-79-6
IUPAC Name
2,4,6-tribromophenol
Molecular Formula
C6H3Br3O
InChI Key
BSWWXRFVMJHFBN-UHFFFAOYSA-N
SMILES
OC1=C(Br)C=C(Br)C=C1Br
Appearance (Color)
White to cream to pink to brown
Appearance (Form)
Powder or crystals or flakes and/or chunks
Infrared spectrum
Conforms
Melting point
87°C to 94°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text