Thermo Scientific Chemicals

2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals

Catalog number: 160120025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals

Catalog number: 160120025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 160120025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-76-4
IUPAC Name
2,4-di-tert-butylphenol
Molecular Formula
C14H22O
InChI Key
ICKWICRCANNIBI-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)C
Appearance (Form)
Crystalline solid
Melting point
54°C to 59°C
GC
>=96.0 %
Water
=<0.1 % (K.F.)
Appearance (Color)
White to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text