Thermo Scientific Chemicals

3,5-Dimethylaniline, 97+%, Thermo Scientific Chemicals

Catalog number: 160195000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Dimethylaniline, 97+%, Thermo Scientific Chemicals

Catalog number: 160195000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 160195000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
108-69-0
IUPAC Name
3,5-dimethylaniline
Molecular Formula
C8H11N
InChI Key
MKARNSWMMBGSHX-UHFFFAOYSA-N
SMILES
CC1=CC(N)=CC(C)=C1
Appearance (Color)
Clear colorless to yellow to brown
Infrared spectrum
Conforms
GC
>=97.0 %
Titration with HClO4
97.0 to 103.0 %
Refractive index
1.5560 to 1.5600 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text