Thermo Scientific Chemicals

1,2,3-Trimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: 160211000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2,3-Trimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: 160211000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 160211000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
634-36-6
IUPAC Name
1,2,3-trimethoxybenzene
Molecular Formula
C9H12O3
InChI Key
CRUILBNAQILVHZ-UHFFFAOYSA-N
SMILES
COC1=CC=CC(OC)=C1OC
Appearance (Form)
Powder or crystals and/or chunks
Melting point
42°C to 47°C
Infrared spectrum
Conforms
Appearance (Color)
White to beige to light brown
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text