Thermo Scientific Chemicals

(-)-Dibenzoyl-L-tartaric acid monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 160251000
100 g, Each
Thermo Scientific Chemicals

(-)-Dibenzoyl-L-tartaric acid monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 160251000
100 g, Each
Quantity
Catalog number: 160251000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62708-56-9
IUPAC Name
(2R,3S)-2,3-bis(benzoyloxy)butanedioic acid
Molecular Formula
C18H14O8
InChI Key
YONLFQNRGZXBBF-OKILXGFUSA-N
SMILES
OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O
Titration with TBAH
97.5 to 102.5 %
Water
4 to 5.5 %
Appearance (Color)
White
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text