Methyl vanillate, 99%, Thermo Scientific Chemicals
Methyl vanillate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl vanillate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 160301000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl 4-hydroxy-3-methoxybenzoate
Molecular FormulaC9H10O4
InChI KeyBVWTXUYLKBHMOX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(O)C(OC)=C1
Molecular Weight (g/mol)182.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or granules
Melting point64°C to 67°C
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only