Thermo Scientific Chemicals

2,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 160320100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 160320100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 160320100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
611-01-8
IUPAC Name
2,4-dimethylbenzoic acid
Molecular Formula
C9H10O2
InChI Key
BKYWPNROPGQIFZ-UHFFFAOYSA-N
SMILES
CC1=CC=C(C(O)=O)C(C)=C1
Infrared spectrum
Conforms
Appearance (Color)
White to beige
Melting point
122°C to 127°C
Titration with NaOH
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text