Hydroxyacetone, Technical, Thermo Scientific Chemicals
Hydroxyacetone, Technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydroxyacetone, Technical, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 160341000
Price (USD)/ Each
42.80
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
42.80
Add to cart
Hydroxyacetone, Technical, Thermo Scientific Chemicals
Catalog number160341000
Price (USD)/ Each
42.80
-
Add to cart
Chemical Identifiers
CAS116-09-6
IUPAC Name1-hydroxypropan-2-one
Molecular FormulaC3H6O2
InChI KeyXLSMFKSTNGKWQX-UHFFFAOYSA-N
SMILESCC(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only