D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals
D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals
D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals
D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals

D(+)-Galactosamine hydrochloride ((2R,3R,4R,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal hydrochloride), CAS # 1772-03-8, is the salt of the amino sugar that acts as an RNA synthesis inhibitor in hepatocytes.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 160440250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1772-03-8
IUPAC Namehydrogen (3R,4R,5R,6R)-3-amino-6-(hydroxymethyl)oxane-2,4,5-triol chloride
Molecular FormulaC6H14ClNO5
InChI KeyQKPLRMLTKYXDST-BMZZJELJSA-N
SMILES[H+].[Cl-].N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Specific optical rotation+93° to +99° (20°C, 589 nm) (c=1, H2O)
Specific optical rotationafter 24 h
Residue after ignition=<1 %
Appearance (Color)White to off-white
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D-Galactosamine is a pyranose amino sugar with hepatotoxic traits

• It acts as an RNA synthesis inhibitor in hepatocytes

• It promotes adherence of polymorphonuclear leukocytes to endothelial cells, increasing superoxide production

Applications

• D-Galactosamine can be used as a hepatotoxic in experimental in vitro models when in combination with LPS

• It is suitable for a range of biochemical laboratory applications

RUO – Research Use Only

General References:

  1. Kennedy, J.F. Selective detection and quantitative determination of the pentoses. 1970, Chromatographia 3, 316–319.
  2. Lloro, I.; Gonzalez, E.; Gutierrez-de Juan, V. et al. Non-invasive detection of drug toxicity in rats by solid-phase extraction and MALDI-TOF analysis of urine samples. Anal Bioanal Chem 2013, 405, 2311–2320.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0446267Certificate of AnalysisMay 17, 2025160440050, 160440010, 160440250
A0413584Certificate of AnalysisDec 13, 2022160440050, 160440010, 160440250
A0375338Certificate of AnalysisDec 13, 2022160440050, 160440010, 160440250
A0412789Certificate of AnalysisDec 13, 2022160440050, 160440010, 160440250
NABSE/TSE CertificationJan 29, 2025160440050, 160440010, 160440250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection