3-Iodobenzoic acid, 98%, Thermo Scientific Chemicals
3-Iodobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Iodobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 160480500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-iodobenzoic acid
Molecular FormulaC7H5IO2
InChI KeyKVBWBCRPWVKFQT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(I)=C1
Molecular Weight (g/mol)248.02
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point185°C to 189°C
HPLC>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only