Thermo Scientific Chemicals

D(-)-Quinic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 160501000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

D(-)-Quinic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 160501000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 160501000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-95-2
IUPAC Name
1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
Molecular Formula
C7H12O6
InChI Key
AAWZDTNXLSGCEK-UHFFFAOYNA-N
SMILES
OC1CC(O)(CC(O)C1O)C(O)=O
Melting point
162°C to 168°C
Titration with NaOH
>=98.0 %
Specific optical rotation
-42° to -45° (20°C, 589 nm) (c=10, water)
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text