Thermo Scientific Chemicals

L(-)-Borneol, 97%, Thermo Scientific Chemicals

Catalog number: 160650250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

L(-)-Borneol, 97%, Thermo Scientific Chemicals

Catalog number: 160650250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 160650250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
464-45-9
IUPAC Name
(1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Molecular Formula
C10H18O
InChI Key
DTGKSKDOIYIVQL-NQMVMOMDSA-N
SMILES
CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
Infrared spectrum
Conforms
Melting point
204°C to 208°C
GC
>=96.0 %
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text