3-Indolebutyric acid, 98%, Thermo Scientific Chemicals
3-Indolebutyric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Indolebutyric acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 160691000
Price (USD)/ Each
402.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
474.00 
Save 71.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
402.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
474.00 
Save 71.35 (15%)
Add to cart
3-Indolebutyric acid, 98%, Thermo Scientific Chemicals
Catalog number160691000
Price (USD)/ Each
402.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
474.00 
Save 71.35 (15%)
-
Add to cart
Chemical Identifiers
CAS133-32-4
IUPAC Name4-(1H-indol-3-yl)butanoic acid
Molecular FormulaC12H13NO2
InChI KeyJTEDVYBZBROSJT-UHFFFAOYSA-N
SMILESOC(=O)CCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point120°C to 125°C
Appearance (Color)White to yellow to beige to brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only