4-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 160721000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1878-66-6
IUPAC Name2-(4-chlorophenyl)acetic acid
Molecular FormulaC8H7ClO2
InChI KeyCDPKJZJVTHSESZ-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 % (K.F.)
Appearance (Color)White to cream
Appearance (Form)Powder
Infrared spectrumConforms
Appearance of solution(5% soln. in CHCl3) clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only