Thermo Scientific Chemicals

4-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 160725000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 160725000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 160725000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1878-66-6
IUPAC Name
2-(4-chlorophenyl)acetic acid
Molecular Formula
C8H7ClO2
InChI Key
CDPKJZJVTHSESZ-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(Cl)C=C1
Water
=<0.2 % (K.F.)
Appearance (Color)
White to cream
Appearance (Form)
Powder
Infrared spectrum
Conforms
Appearance of solution
(5% soln. in CHCl3) clear

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text