4-Fluorobenzonitrile, 99%
4-Fluorobenzonitrile, 99%
4-Fluorobenzonitrile, 99%
4-Fluorobenzonitrile, 99%
Thermo Scientific Chemicals

4-Fluorobenzonitrile, 99%

CAS: 1194-02-1 | C7H4FN | 121.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 160810050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1194-02-1
IUPAC Name4-fluorobenzonitrile
Molecular FormulaC7H4FN
InChI KeyAEKVBBNGWBBYLL-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Low melting crystalline solid
Melting point33°C to 36°C
GC>=98.5 %
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link