Thermo Scientific Chemicals

2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 160902500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 160902500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 160902500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-84-0
IUPAC Name
2,4-dichlorobenzoic acid
Molecular Formula
C7H4Cl2O2
InChI Key
ATCRIUVQKHMXSH-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(Cl)C=C1Cl
Appearance (Color)
White to off-white
Titration with NaOH
>=97.5 %
HPLC
>=97.5 %
Water
=<0.5 % (K.F.)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text