Methyl 2,2,2-trichloroacetimidate, 98%, Thermo Scientific Chemicals
Methyl 2,2,2-trichloroacetimidate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2,2,2-trichloroacetimidate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 160930250
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
121.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
121.00 
Save 18.35 (15%)
Add to cart
Methyl 2,2,2-trichloroacetimidate, 98%, Thermo Scientific Chemicals
Catalog number160930250
Price (USD)/ Each
102.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
121.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2533-69-9
IUPAC Namemethyl 2,2,2-trichloroethanimidate
Molecular FormulaC3H4Cl3NO
InChI KeyOGBINJLTBZWRRB-UHFFFAOYSA-N
SMILESCOC(=N)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4770 to 1.4800 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only