Thermo Scientific Chemicals

Benzoyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 160951000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Benzoyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 160951000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 160951000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
618-32-6
IUPAC Name
benzoyl bromide
Molecular Formula
C7H5BrO
InChI Key
AQIHMSVIAGNIDM-UHFFFAOYSA-N
SMILES
BrC(=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to light orange
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.5880 to 1.5940 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text