Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 160971000
Price (USD)/ Each
60.65
Online exclusive
67.70 
Save 7.05 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
67.70 
Save 7.05 (10%)
Add to cart
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Catalog number160971000
Price (USD)/ Each
60.65
Online exclusive
67.70 
Save 7.05 (10%)
-
Add to cart
Chemical Identifiers
CAS103-72-0
IUPAC Nameisothiocyanatobenzene
Molecular FormulaC7H5NS
InChI KeyQKFJKGMPGYROCL-UHFFFAOYSA-N
SMILESS=C=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear to hazy colorless to yellow to orange
AppearanceMay develop some turbidity or precipitate
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only