Pierce™ Glutathione Agarose
Pierce™ Glutathione Agarose
Thermo Scientific™

Pierce™ Glutathione Agarose

Thermo Scientific Pierce Glutathione Agarose is a high-capacity, high-performance resin for affinity purification of GST-tagged fusion proteins from cellular lysates.FeaturesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
16101100 mL
16102BID500 mL
1610010 mL
Catalog number 16101
Price (USD)
1,560.65
Online Exclusive
1,746.00
Save 185.35 (11%)
Each
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)
1,560.65
Online Exclusive
1,746.00
Save 185.35 (11%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce Glutathione Agarose is a high-capacity, high-performance resin for affinity purification of GST-tagged fusion proteins from cellular lysates.

Features of Glutathione Agarose:

Slurry—crosslinked 6% beaded agarose, slurried in water with sodium azide
High capacity—binds at least 40 mg of purified recombinant GST protein per milliliter of resin
High yield and purity—consistently purifies at least 10 mg of GST-tagged protein per milliliter of resin with greater than 90% purity
Cost-effective—resin is economically priced and can be reused at least five times without reduction in binding capacity and purification performance
Versatile—works well to purify GST-fusion proteins from bacterial lysates or use with pre-purified GST-tagged proteins to pull down protein interactions
Compatible—validated and effective for use with Thermo Scientific Cell Lysis Reagents to extract and purify from bacterial or mammalian cell cultures

The high-quality support consists of glutathione that has been immobilized by its central sulfhydryl group via a 12-atom spacer arm to crosslinked 6% beaded agarose. Purification of GST-fusion proteins using glutathione (GSH) agarose beads is well documented and adaptable to a variety of scales, column formats and specific applications. Whether the purpose is to purify large amounts of recombinant protein from over-expressing E. coli lysates or to investigate protein interactions involving GST-tagged bait proteins, Pierce Glutathione Agarose is suitable for the task.

Pierce Glutathione Agarose effectively purifies high levels of overexpressed GST-tagged fusion proteins from bacterial lysates, such as those that are obtained with Thermo Scientific B-PER Bacterial Protein Extraction Reagents.

Pierce Glutathione Agarose performs well in batch-binding and spin-column procedures at a variety of scales. Performance equals or exceeds popular GSH resins from other suppliers.

Pierce Glutathione Agarose is a high-quality, stable and resilient affinity support. Tests confirm that no decrease in performance occurs after at least five repeated uses. These data indicated that the resin is highly resistant to structural degradation or ligand leaching during normal use.

The expression and purification of recombinant proteins is central to protein regulation, structure and function studies. The majority of recombinant proteins are expressed as fusions with short affinity tags or small proteins, such as glutathione S-transferase (GST). This protein binds specifically to reduced glutathione (GSH) in near-neutral, nondenaturing conditions (e.g., Tris buffer). Bound protein is easily dissociated (eluted) by competitive displacement with buffer containing free, reduced GSH (oxidized glutathione, GSSH is not effective for this purpose). When proteins of interest are expressed as fusions with GST and glutathione is immobilized to an solid support, this protein-substrate system enables affinity purification of recombinant proteins, as well as various other experiments with those proteins.

Related Products
Pierce™ Glutathione Spin Columns, 0.2 mL
Pierce™ Glutathione Spin Columns, 1 mL
Pierce™ Glutathione Spin Columns, 3 mL
Pierce™ Glutathione Chromatography Cartridges, 1 mL
Pierce™ Glutathione Superflow Agarose
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeAffinity, Agarose Resin
FormatBottle
Protein FormRecombinant
Quantity100 mL
Stationary PhaseGlutathione
TargetGST-Tagged Protein
Capacity (Metric)40 mg Purified Recombinant GST Protein Binding per mL Resin
FormLiquid
Product LinePierce™
TypeAgarose
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.
croppedsalg3fam

Slide-A-Lyzer  G3 Dialysis Cassettes
Make your dialysis simpler and more secure

Learn more ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link