3-Methoxyphenol, 97%, Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxyphenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 161060250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-methoxyphenol
Molecular FormulaC7H8O2
InChI KeyASHGTJPOSUFTGB-UHFFFAOYSA-N
SMILESCOC1=CC=CC(O)=C1
Molecular Weight (g/mol)124.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pink-red to brown
Appearance (Form)Liquid
Refractive index1.5510 to 1.5530 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only