Thermo Scientific Chemicals

3-Methoxyphenol, 97%, Thermo Scientific Chemicals

Catalog number: 161065000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Methoxyphenol, 97%, Thermo Scientific Chemicals

Catalog number: 161065000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 161065000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
150-19-6
IUPAC Name
3-methoxyphenol
Molecular Formula
C7H8O2
InChI Key
ASHGTJPOSUFTGB-UHFFFAOYSA-N
SMILES
COC1=CC=CC(O)=C1
Appearance (Color)
Clear pink-red to brown
Appearance (Form)
Liquid
Refractive index
1.5510 to 1.5530 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text