Thermo Scientific™

Pierce™ Glutathione Chromatography Cartridges, 1 mL

Catalog number: 16109
Have Questions?
Thermo Scientific™

Pierce™ Glutathione Chromatography Cartridges, 1 mL

Catalog number: 16109
Have Questions?
Catalog Number
16109
Unit Size
5 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
161095 x 1 mL
-
161102 x 5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Glutathione Chromatography Cartridges are convenient, ready-to-use pre-packed devices for thepurification of GST-fusion proteins from cellular lysates.

Features of Glutathione Agarose:

High capacity—binds at least 40 mg of purified recombinant GST protein per milliliter of resin
High yield and purity—consistently purifies at least 10 mg of GST-tagged protein per milliliter of resin with greater than 90% purity
Cost-effective—resin is economically priced and can be reused at least five times without reduction in binding capacity and purification performance
Versatile—works well to purify GST-fusion proteins from bacterial lysates or use with pre-purified GST-tagged proteins to pull down protein interactions
Compatible—validated and effective for use with Thermo Scientific Cell Lysis Reagents to extract and purify from bacterial or mammalian cell cultures

The glutathione is immobilized through its central sulfhydryl onto 6% crosslinked agarose resin. Purification of GST-fusion proteins using glutathione (GSH) agarose beads is well documented and provides a one-step, high purity affinity purification. Whether the purpose is to purify large amounts of recombinant protein from over-expressing E. coli lysates or to investigate protein interactions involving GST-tagged bait proteins, Pierce Glutathione Agarose is suitable for the task.

Thermo Scientific Pierce Glutathione Agarose is also offered in three volumes of resin slurry, complete GST purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs.

Pierce Glutathione Agarose effectively purifies high levels of overexpressed GST-tagged fusion proteins from bacterial lysates, such as those that are obtained with Thermo Scientific B-PER Bacterial Protein Extraction Reagents.

Pierce Glutathione Agarose performs well in batch-binding and spin-column procedures at a variety of scales. Performance equals or exceeds popular GSH resins from other suppliers.

Pierce Glutathione Agarose is a high-quality, stable and resilient affinity support. Tests confirm that no decrease in performance occurs after at least five repeated uses. These data indicated that the resin is highly resistant to structural degradation or ligand leaching during normal use.

The expression and purification of recombinant proteins is central to protein regulation, structure and function studies. The majority of recombinant proteins are expressed as fusions with short affinity tags or small proteins, such as glutathione S-transferase (GST). This protein binds specifically to reduced glutathione (GSH) in near-neutral, nondenaturing conditions (e.g., Tris buffer). Bound protein is easily dissociated (eluted) by competitive displacement with buffer containing free, reduced GSH (oxidized glutathione, GSSH is not effective for this purpose). When proteins of interest are expressed as fusions with GST and glutathione is immobilized to an solid support, this protein-substrate system enables affinity purification of recombinant proteins, as well as various other experiments with those proteins.

Related Products
Pierce™ Glutathione Chromatography Cartridges, 1 mL
Pierce™ Glutathione Spin Columns, 3 mL
Pierce™ Glutathione Agarose
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
1 mL
Description
Pierce Glutathione Chromatography Cartridges, 1mL
Form
Liquid
Product Line
Pierce™
Quantity
5 Cartridges
Protein Form
Recombinant
Target
GST-Tagged Protein
Type
Chromatography Cartridge
Stationary Phase
Glutathione
Column Type
Affinity, Agarose Resin

Contents & Storage

Upon receipt store at 4–8°C. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text