Chicken Serum, New Zealand Origin
Chicken Serum, New Zealand Origin
Gibco™

Chicken Serum, New Zealand Origin

Sustainable packaging
Have Questions?
Catalog NumberQuantity
16110082500 mL
Catalog number 16110082
Price (USD)
130.00
Each
-
Add to cart
Quantity:
500 mL
Price (USD)
130.00
Each
Add to cart
Ask our AI about this Product

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Specialty
  • Origin: New Zealand
  • Age: Adult

Cell Culture, Mammalian Cell Culture

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Product LineGibco™
Shipping ConditionFrozen
SpeciesChicken
AgeAdult
Country of OriginNew Zealand
FormLiquid
Product TypeChicken Serum
Quantity500 mL
Serum TreatmentSterile-filtered
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection