Thermo Scientific Chemicals

Veratrole, 99+%, Thermo Scientific Chemicals

Catalog number: 161115000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Veratrole, 99+%, Thermo Scientific Chemicals

Catalog number: 161115000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 161115000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
91-16-7
IUPAC Name
1,2-dimethoxybenzene
Molecular Formula
C8H10O2
InChI Key
ABDKAPXRBAPSQN-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1OC
Appearance (Form)
Clear liquid or solid
Infrared spectrum
Conforms
GC
>=99.0 %
Color scale
=<50 APHA
Refractive index
1.5320 to 1.5360 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text