Thermo Scientific™

Active Cdc42 Pull-Down and Detection Kit

Catalog number: 16119
Have Questions?
Thermo Scientific™

Active Cdc42 Pull-Down and Detection Kit

Catalog number: 16119
Have Questions?
Catalog Number
16119
Unit Size
30 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1611930 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Active Cdc42 Pull-Down and Detection Kit is a complete kit for selective enrichment and detection of GTP-bound Cdc42 GTPase through specific protein interaction with the Pak1 protein-binding domain.

The Active Cdc42 Pull-Down and Detection Kit includes purified GST-Pak1 protein-binding domain (PBD), glutathione agarose resin, positive and negative controls (GTPγS and GDP, respectively), lysis/binding/wash buffer, anti-Cdc42 primary antibody, sample buffer, spin columns and collection tubes. The kit was validated using lysates from NIH 3T3 cells, a cell line that is known to have robust Cdc42 activity.

Features of the Active Cdc42 Pull-Down and Detection Kit:

Highly sensitive and accurate—optimized reagents, specific anti-Cdc42 antibody and Western blot procedure ensure accurate controls and semi-quantitative results
Validated—functionally tested for Cdc42 detection to ensure quality and performance
Compatible—effective with a variety of cell types from mouse, rat and human sources

Applications:
• Follow activation of Cdc42 GTPase during cell differentiation, migration, division, and cytoskeletal rearrangement
• Study the activation of Cdc42 during filopodia formation
• Monitor Cdc42 activity after stimulation with growth factors
• Screen small molecule inhibitors for their effects on Cdc42 activity

The Active Cdc42 Pull-Down and Detection Kit was validated for the function and specificity of the active Cdc42 enrichment method using cell lysates treated with GTPγS to activate endogenous Cdc42 and compared to lysates treated with GDP to inactivate the small GTPase. GTPγS treatment traps Cdc42 in the GTP-bound, active form, resulting in a strong signal when endogenous Cdc42 is present. GDP treatment pushes Cdc42 into the GDP-bound, inactive state, resulting in minimal or no signal regardless of Cdc42 protein levels. This kit is optimized for Western blot detection with an HRP-conjugated secondary antibody (Goat Anti-mouse IgG, Part No. 31430) and Thermo Scientific SuperSignal West Pico Chemiluminescent Substrate (Part No. 34080). The kit contains sufficient components for 30 pull-down assays.

Cdc42 Background:
Rho family GTPases serve many cellular functions, including cell signaling, transcriptional regulation and organization of the actin cytoskeleton. This family of GTPases comprise Rho (RhoA, RhoB, and RhoC), Rac (Rac1, Rac2, Rac3, and RhoG), Cdc42 (Cdc42 and G25K), Rnd (Rnd1, Rnd2, and RhoE/Rnd3), RhoBTB family and the Miro family. These GTPases enable signal transduction from the plasma membrane to the cytosol through GPCR, tyrosine kinase, cytokine and adhesion receptors. Attachment to the plasma membrane is accomplished through geranylgeranyl lipid modifications at the carboxy-terminus of the protein. Cdc42 activation results in the polymerization of actin filaments and filopodia formation.

The Cdc42 subfamily of RhoGTPase has been less well-characterized than the Rho and Rac families. Both Cdc42 and Rac1 are activated through tyrosine receptor kinase signaling leading to SAPK and p38 stress kinase pathway activation. Cdc42 is also activated by the chemoattractant fMLP in neutrophils. Fibronectin activates Cdc42 and Rac1 to induce cell spreading, and stimulation with TNF-alpha and IL-1 results in changes in the actin cytoskeleton. There is significant cross-talk between Cdc42 and Rac1, as they act in overlapping pathways, and in some cases, Cdc42 may act upstream of Rac1 during signal transduction. Some of the main effector proteins of Cdc42 are Pak1, N-WASP and IQGAP. Pak1 is a kinase involved in the activation of JNK in the SAPK stress pathway. N-WASP is an effector that induces filopodia formation, and IQGAP interacts with F-actin filaments. In differentiating neurons, Cdc42 plays an active role in neurite outgrowth. However, the Rho family of GTPases can work agonistically during cell signaling and antagonistically during differentiation.

More Product Data
Measure activation of small GTPases via their specific downstream effectors

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Pierce Active Cdc42 Pull-Down and Detection Kit
Assay
Protein Quantitation Assay
Conjugate
HRP
Detection Method
Western Blot, Chemiluminescent
Immunoassay Kit Format
Spin Column
Label Type
Unlabeled
Substrate Properties
Protein-Based Substrate
Substrate Type
GTPase Substrate
For Use With (Application)
Pull-Down
Kit Contents
Anti-Cdc42 Antibody, 5 units, 300µL, Collection Tubes, 90 tubes, GDP (100X), 50µL, Glutathione Agarose Resin, 3mL, GST-Pak1-RBD Fusion Protein, 1 vial, GTPγS (100X), 50μL, Lysis/Binding/Wash Buffer (1X), 100mL, SDS Sample Buffer (2X), 1.5mL, Spin Cups, 30 cups
Target
Kinases (Multiple), Phosphodiesterases, ATPases, GTPases, Phosphatases

Contents & Storage

• GST-Pak1-RBD Fusion Protein, 1 vial 
• Anti-Cdc42 Antibody, 5 units, 300 μL 
• Glutathione Agarose Resin, 3 mL 
• GDP (100X), 50 μL 
• GTPyS (100X), 50 μL 
• SDS Sample Buffer (2X), 1.5 mL 
• Lysis/Binding/Wash Buffer (1X), 100 mL 
• Spin Cups, 30 cups 
• Collection Tubes, 90 tubes 

See product manual for storage conditions of individual components.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text