Decanoyl chloride, 98+%, Thermo Scientific Chemicals
Decanoyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decanoyl chloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 161191000
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.20 
Save 6.55 (14%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.20 
Save 6.55 (14%)
Add to cart
Decanoyl chloride, 98+%, Thermo Scientific Chemicals
Catalog number161191000
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.20 
Save 6.55 (14%)
-
Add to cart
Chemical Identifiers
CAS112-13-0
IUPAC Namedecanoyl chloride
Molecular FormulaC10H19ClO
InChI KeyIPIVAXLHTVNRBS-UHFFFAOYSA-N
SMILESCCCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4400 to 1.4430 (20°C, 589 nm)
Color scale=<50 APHA
GC>=98.0 %
Carboxylic acid=<2 % (as Decanoic acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only