Thermo Scientific Chemicals

Decanoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 161195000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Decanoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: 161195000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 161195000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-13-0
IUPAC Name
decanoyl chloride
Molecular Formula
C10H19ClO
InChI Key
IPIVAXLHTVNRBS-UHFFFAOYSA-N
SMILES
CCCCCCCCCC(Cl)=O
Infrared spectrum
Conforms
Refractive index
1.4400 to 1.4430 (20°C, 589 nm)
Color scale
=<50 APHA
GC
>=98.0 %
Carboxylic acid
=<2 % (as Decanoic acid)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text