Thermo Scientific Chemicals

N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals

Catalog number: 161255000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals

Catalog number: 161255000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 161255000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4637-24-5
IUPAC Name
(dimethoxymethyl)dimethylazanium
Molecular Formula
C5H14NO2
InChI Key
ZSXGLVDWWRXATF-UHFFFAOYSA-O
SMILES
COC(OC)[NH+](C)C
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.3950 to 1.3970 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text