Thermo Scientific Chemicals

Tetrabutylammonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: 161270025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: 161270025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 161270025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
311-28-4
IUPAC Name
tetrabutylazanium iodide
Molecular Formula
C16H36IN
InChI Key
DPKBAXPHAYBPRL-UHFFFAOYSA-M
SMILES
[I-].CCCC[N+](CCCC)(CCCC)CCCC
Appearance (Color)
White to cream
Appearance (Form)
Powder or crystals or flakes
Infrared spectrum
Conforms
Melting point
141°C to 150°C
Titration Argentometric
>=97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text