Thermo Scientific Chemicals

2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog number: 161340010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog number: 161340010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 161340010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4733-39-5
IUPAC Name
2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
Molecular Formula
C26H20N2
InChI Key
STTGYIUESPWXOW-UHFFFAOYSA-N
SMILES
CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1
Appearance (Color)
White to cream to yellow
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
277°C to 285°C (Dec)
Titration with HClO4
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text