D(-)-Fructose, 99%
Thermo Scientific Chemicals

D(-)-Fructose, 99%

D(-)-Fructose, 99%, C6H12O6, CAS Number-57-48-7, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, arabino-hexulose, d--fructose, d---fructose, d-levulose, fructose, d, furucton, keto-d-fructose, krystar 300, sugar, fruit, 500g, Passes test, 98.5% min, 31,321, CHEBI:48095, 50ppm max, White, 200-333-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 161350025
Price (USD)/ Each
331.65
Online exclusive
368.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
331.65
Online exclusive
368.00 
Save 36.35 (10%)
Add to cart
D(-)-Fructose, 99%
Catalog number161350025
Price (USD)/ Each
331.65
Online exclusive
368.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS57-48-7
IUPAC Name(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Molecular FormulaC6H12O6
InChI KeyBJHIKXHVCXFQLS-UYFOZJQFSA-N
SMILESOC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Chloride (Cl)=<50 ppm
Infrared spectrumConforms
Appearance (Color)White
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only