Thermo Scientific™

EZ-Link™ Desthiobiotinylation and Pull-Down Kit

Catalog number: 16138
Have Questions?
Thermo Scientific™

EZ-Link™ Desthiobiotinylation and Pull-Down Kit

Catalog number: 16138
Have Questions?
Catalog Number
16138
Unit Size
5 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
161385 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific EZ-Link Desthiobiotinylation and Pull-Down Kit is a complete set of reagents to biotin-label purified proteins and use them to purify interaction complexes using streptavidin agarose resin with mild elution conditions.

Features of the EZ-Link Desthiobiotinylation and Pull-Down Kit:

Sulfo-NHS-LC-Desthiobiotin – label proteins with a biotin-like that specifically binds streptavidin but also easily elutes with mild conditions
Pull-down assay – simple, optimized protocol and reagents to label proteins and recover (purify) intact, functional protein interaction complexes
Complete kit – includes labeling reagent and desalting columns to prepare bait-proteins, and streptavidin agarose and elution buffer to perform pull-down

This all-in-one labeling and purification kit uses Sulfo-NHS-LC-Desthiobiotin, an amine-reactive NHS-ester reagent, to effectively tag a purified or mixed protein sample for use as bait in capturing and purifying protein interactions. The desthiobiotin tags of labeled proteins bind to streptavidin with high specificity yet readily elute with very mild conditions (i.e., by competitive displacement with regular, free biotin). The system enables effective yet gentle capture and isolation (pull-down) of specific protein interaction complexes from cell lysates using streptavidin agarose beads. The complete kit includes ready-to-use 1 mg vials of desthiobiotinylation reagent, desalting columns to process labeled protein, and high-capacity streptavidin agarose with buffers to perform several microcentrifuge-scale pull-down assay experiments with ease. Identify novel interactions between a known protein (bait) and previously undiscovered target (prey), or confirm interactions among known bait and prey proteins.

Desthiobiotin is a single-ring, sulfur-free analog of biotin that binds to streptavidin with nearly equal specificity but less affinity than biotin (1/Kd = 1011 vs. 1015M, respectively). Consequently, desthiobiotinylated bait proteins and their interacting partners can be eluted readily and specifically from streptavidin affinity resin using mild conditions based on competitive displacement with free biotin. For pull-down assay experiments with biological samples, this soft-release characteristic of desthiobiotin also helps to minimize co-purification of endogenous biotinylated molecules, which remain bound to streptavidin upon elution of the target protein complexes with free biotin. The modified avidin-biotin affinity system also eliminates the need to use harsh elution conditions that might disassociate complexes and/or damage the target protein or cell. Desthiobiotin-based techniques are ideal when using native or recombinant proteins that are not expressed with a fusion tag and when isolating captured proteins under native conditions, such as targeting intact cells or cell surface proteins.

Sulfo-NHS-LC-Desthiobiotin is a variant of biotin that is activated as a sulfo-NHS ester with a long-chain (LC) spacer arm to covalently label primary amines (-NH2) of proteins or other molecules with desthiobiotin groups. The No-Weigh Format (1 mg resealable vials) eliminates the difficulties associated with weighing small quantities of reagent and maximizes protection of unused reagent from hydrolysis.

Related Products
Pierce™ Biotinylated Protein Interaction Pull-Down Kit
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
Affinity Purification
For Use With (Equipment)
Microcentrifuge
Product Line
EZ-Link™
Quantity
1 Kit
For Use With (Application)
Pull-Down (Biotin-Based)
Target
Not Target-Specific
Type
Pull-Down Kit
Sufficient For
5 Reactions
Format
Kit

Contents & Storage

Sufficient For: 5 labeling reactions (0.2-5 mg protein each) and up to 40 pull-down experiments (50 μL each)
• EZ-Link Sulfo-NHS-LC-Desthiobiotin, 2 x 1 mg
• Zeba Spin Desalting Column (7K MWCO), 5 mL, 5 columns
• High Capacity Streptavidin Agarose Resin, 2 mL
• PBS Pack (makes 500mL), 1 pack
• Biotin Elution Buffer, 8 mL

Store EZ-Link Sulfo-NHS-LC-Desthiobiotin at -20°C. Store PBS solution at room temperature or 4°C once reconstituted. Store remaining kit components at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text