trans-Ethyl crotonate, 96%, Thermo Scientific Chemicals
trans-Ethyl crotonate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-Ethyl crotonate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 161385000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS623-70-1
IUPAC Nameethyl (2E)-but-2-enoate
Molecular FormulaC6H10O2
InChI KeyZFDIRQKJPRINOQ-HWKANZROSA-N
SMILESCCOC(=O)\C=C\C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4220 to 1.4280 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=95 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only