Informational:Informational:100% knock-in using CRISPR-Cas9—try our new TrueTag Donor DNA Kits for fluorescent or epitope gene tagging.
Thermo Scientific™

pCMV-Gaussia Luc Vector for Luciferase Assays

Catalog number: 16147
Have Questions?
Thermo Scientific™

pCMV-Gaussia Luc Vector for Luciferase Assays

Catalog number: 16147
Have Questions?
Catalog Number
16147
Unit Size
10 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1614710 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific pCMV-Gaussia Luc vector is a derivative of the pMCS-Gaussia Luc vector containing the Gaussia luciferase gene under the control of the CMV (Cytomegalovirus) promoter. This constitutive expression vector can be used as a normalization control to account for experimental variation in combination with other reporters.

Features of the pCMV-Gaussia Luc vector:

• Strong (CMV) constitutive promoters for co-transfection and normalization
• Naturally-secreting Gaussia luciferase gene, optimized for high expression in mammalian systems
• Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
• Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
• Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
• High-copy pUC bacterial DNA replication origin

The Gaussia Luc Vectors contain a gene cloned from the copepod, Gaussia princeps. The gene encodes a naturally-secreted bioluminescent Gaussia luciferase (approx. 20kDa), which enables measurement of the reporter activity in media (for real-time assays) and in cell lysates.

These vectors are subject to a limited use label license.

More Product Data
Highly sensitive multiplex luciferase reporter assays
Luciferase assays in hard-to-transfect Jurkat cells
Monitoring neuronal differentiation using multiplexed luciferase reporters
Activation of the antioxidant response pathway by pesticide chemicals

Related Products
pMCS-Gaussia Luc Vector for Luciferase Assays
pTK-Gaussia Luc Vector for Luciferase Assays
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
pCMV-Gaussia Luc Vector
Type
Luc Vector
Cloning Method
Restriction Enzyme/MCS
Delivery Type
Transfection
Quantity
10 μg
Reporter Gene
Gaussia Luciferase
Selection Agent (Eukaryotic)
Puromycin
For Use With (Application)
Reporter Assays
Promoter
CMV

Contents & Storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: -70 to -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text