Thermo Scientific Chemicals

2-chloro-6-fluorobenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 161510250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-chloro-6-fluorobenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 161510250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 161510250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
387-45-1
IUPAC Name
2-chloro-6-fluorobenzaldehyde
Molecular Formula
C7H4ClFO
InChI Key
OACPOWYLLGHGCR-UHFFFAOYSA-N
SMILES
FC1=CC=CC(Cl)=C1C=O
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline solid
GC
>=94 %
Melting point
34°C to 39°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text