Thermo Scientific™
pMCS-Green Renilla Luc Vector for Luciferase Assays
Catalog number: 16152
Have Questions?
Thermo Scientific™
pMCS-Green Renilla Luc Vector for Luciferase Assays
Catalog number: 16152
Have Questions?
Catalog Number
16152
Unit Size
10 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1615210 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific pMCS-Renilla Luc vector is a multiple cloning site plasmid to designed to accept a promoter sequence for study of gene regulation using the intracellular Renilla luciferase reporter.
Features of the pMCS-Renilla Luc vector:
• Intracellular Green Renilla luciferase gene, optimized for high expression in mammalian systems
• Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
• Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
• Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
• High-copy pUC bacterial DNA replication origin
The Green Renilla Luc Vectors contain a gene cloned from the sea pansy, Renilla reniformis. The gene encodes a bioluminescent Green Renilla luciferase (36kDa). The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential.
These vectors are subject to a limited use label license.
More Product Data
Luciferase assays in hard-to-transfect Jurkat cells
Related Products
pCMV-Green Renilla Luc Vector for Luciferase Assays
pTK-Green Renilla Luc Vector for Luciferase Assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
pMCS-Green Renilla Luc Vector
Type
Luc Vector
Cloning Method
Restriction Enzyme/MCS
Delivery Type
Transfection
Quantity
10 μg
Reporter Gene
Green Renilla Luciferase
Selection Agent (Eukaryotic)
Puromycin
Product Line
Pierce™
For Use With (Application)
Reporter Assays
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: -70 to -20°C.