Thermo Scientific™
pCMV-Green Renilla Luc Vector for Luciferase Assays
Catalog number: 16153
Have Questions?
Thermo Scientific™
pCMV-Green Renilla Luc Vector for Luciferase Assays
Catalog number: 16153
Have Questions?
Catalog Number
16153
Unit Size
10 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1615310 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific pCMV-Green Renilla Luc vector is a derivative of pMCS-Green Renilla Luc. It contains the luciferase gene under the control of the CMV (Cytomegalovirus) promoter, creating a constitutive expression vector that can be used as a normalization control to account for experimental variation in combination with other reporters.
Features of the pCMV-Green Renilla Luc vector:
• Strong (CMV) constitutive promoter for co-transfection and normalization
• Intracellular Green Renilla luciferase gene, optimized for high expression in mammalian systems
• Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
• Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
• High-copy pUC bacterial DNA replication origin
More Product Data
Luciferase assays in hard-to-transfect Jurkat cells
Related Products
pMCS-Green Renilla Luc Vector for Luciferase Assays
pTK-Green Renilla Luc Vector for Luciferase Assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
pCMV-Green Renilla Luc Vector
Type
Luc Vector
Cloning Method
Restriction Enzyme/MCS
Delivery Type
Transfection
Quantity
10 μg
Reporter Gene
Green Renilla Luciferase
Selection Agent (Eukaryotic)
Puromycin
Product Line
Pierce™
For Use With (Application)
Reporter Assays
Promoter
CMV
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: -70 to -20°C.